Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE6G Antibody (OTI2F5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273298H
Description
PDE6G Monoclonal specifically detects PDE6G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PDE6G | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PDE6G | |
Full length recombinant protein of human PDE6G (NP_002593) produced in HEK293T cells. | |
0.1 mL | |
5148.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2F5 | |
Western Blot, Immunohistochemistry | |
DKFZp686C0587, GMP-PDE gamma, MGC125749, PDEG, phosphodiesterase 6G, cGMP-specific, rod, gamma, retinal rod rhodopsin-sensitive cGMP 3′-5′-cyclic phosphodiesterase subunit gamma, rod cG-PDE G, RP57 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction