Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE6G Monoclonal Antibody (OTI1F3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503686S
Description
PDE6G Monoclonal antibody specifically detects PDE6G in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PDE6G | |
| Monoclonal | |
| 0.74 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| GMP-PDE gamma; p3AT; PDE6G; PDEG; phosphodiesterase 6G; phosphodiesterase 6G, cGMP-specific, rod, gamma; phosphodiesterase, cyclic GMP (rod receptor), gamma polypeptide; retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma; rod cG-PDE G; RP57; unnamed protein product | |
| Full length human recombinant protein of human KIAA0495 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI1F3 | |
| Unconjugated | |
| TP73-AS1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57212 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction