Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE6H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583757 Supplier Invitrogen™ Supplier No. PA583757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRKGPPKFKQ RQTRQFKSKP PKKGVKGFGD DIPGMEGLGT DITVICPWEA FSHLELHELA Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Participates in processes of transmission and amplification of the visual signal. cGMP-PDEs are the effector molecules in G-protein-mediated phototransduction in vertebrate rods and cones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6H
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDE6H
Gene Accession No. P61250, Q13956
Gene Alias A930033D18Rik; ACHM6; cone phosphodiesterase 6 gamma; cone-like cGMP-phosphodiesterase 6 gamma subunit type 2; GMP-PDE gamma; Pde6g; Pde6h; PDEgamma; phosphodiesterase 6G, cGMP-specific, rod, gamma; phosphodiesterase 6H; phosphodiesterase 6H, cGMP-specific, cone, gamma; RCD3; retinal cone rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma
Gene Symbols PDE6H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDE6H. Recombinant protein control fragment (Product #RP-108364).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114248, 5149
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.