Learn More
Invitrogen™ PDE7A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532975
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Bovine, Dog, Bat, Elephant, Panda, Rabbit, Opossum, Xenopus (100%); Mouse, Rat, Hamster, Turkey, Lizard (94%); Zebrafish (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE7 subfamily. This PDE hydrolyzes the second messenger, cAMP, which is a regulator and mediator of a number of cellular responses to extracellular signals. Thus, by regulating the cellular concentration of cAMP, this protein plays a key role in many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
PDE7A | |
Polyclonal | |
Unconjugated | |
PDE7A | |
AU015378; AW047537; HCP1; High affinity cAMP-specific 3',5'-cyclic phosphodiesterase 7A; P2A; PDE7; PDE7-1; Pde7a; phosphodiesterase 7A; phosphodiesterase isozyme 7; rolipram-insensitive phosphodiesterase type 7; TM22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100352319, 100849042, 486975, 5150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 0.1% sodium azide | |
Q13946 | |
PDE7A | |
Synthetic 17 amino acid peptide from N-terminus of human PDE7A. | |
50 μg | |
Primary | |
Human, Canine, Rabbit, Monkey, Bovine, Frog, Bat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.