Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE7A Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208406
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8406 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8406 Supplier Fabgennix Inc Supplier No. PD7A112AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE7A Polyclonal antibody specifically detects PDE7A in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen PDE7A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PDE7A
Gene Accession No. O08593, P70453, Q13946
Gene Alias AU015378; AW047537; HCP1; High affinity cAMP-specific 3',5'-cyclic phosphodiesterase 7A; P2A; PDE7; PDE7-1; Pde7a; phosphodiesterase 7A; phosphodiesterase isozyme 7; rolipram-insensitive phosphodiesterase type 7; TM22
Host Species Rabbit
Immunogen Synthetic cyclic peptide common to PDE7A1 and PDE7A2 variants.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18583, 5150, 81744
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.