Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE7B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152979
Description
PDE7B Polyclonal specifically detects PDE7B in Human samples. It is validated for Western Blot.Specifications
PDE7B | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
bA472E5.1, cAMP-specific 3'-5'-cyclic phosphodiesterase 7B, EC 3.1.4, EC 3.1.4.17, high-affinity cAMP-specific 3'-5'-cyclic phosphodiesterase, MGC88256, phosphodiesterase 7B, rolipram-insensitive phosphodiesterase type 7 | |
Rabbit | |
52 kDa | |
100 μL | |
Signal Transduction, Vision | |
27115 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9NP56 | |
PDE7B | |
Synthetic peptides corresponding to PDE7B(phosphodiesterase 7B) The peptide sequence was selected from the middle region of PDE7B. Peptide sequence IGMLRESRLLAHLPKEMTQDIEQQLGSLILATDINRQNEFLTRLKAHLHN. | |
Affinity purified | |
RUO | |
Primary | |
Expected to cross react based on sequence identity: Zebrafish: 75%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction