Learn More
Invitrogen™ PDE7B Polyclonal Antibody

Description
Recommended positive controls: Rat tissues.
Specifications
Specifications
| Antigen | PDE7B |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Tris glycine with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.9 |
| Gene | Pde7b |
| Gene Accession No. | Q9NP56 |
| Gene Alias | bA472E5.1; cAMP-specific 3',5'-cyclic phosphodiesterase 7B; high-affinity cAMP-specific 3',5'-cyclic phosphodiesterase; MGC88256; Pde7b; phosphodiesterase 7B; rolipram-insensitive phosphodiesterase type 7; RP11-472E5.2 |
| Gene Symbols | Pde7b |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.