Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE7B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981486
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1486 Supplier Bioss Supplier No. BS11576R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The 3′, 5′-cyclic nucleotides cAMP and cGMP function as second messengers in a wide variety of signal transduction pathways. 3′, 5′-cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and cGMP to the corresponding 5′-monophosphates and provide a mechanism to downregulate cAMP and cGMP signaling. This gene encodes a cAMP-specific phosphodiesterase, a member of the cyclic nucleotide phosphodiesterase family.

Specifications

Antigen PDE7B
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Pde7b
Gene Accession No. Q9NP56
Gene Alias bA472E5.1; cAMP-specific 3′,5′-cyclic phosphodiesterase 7B; high-affinity cAMP-specific 3′,5′-cyclic phosphodiesterase; MGC88256; PDE7B; phosphodiesterase 7B; rolipram-insensitive phosphodiesterase type 7; RP11-472E5.2
Gene Symbols Pde7b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PDE7B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27115
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.