Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE8A Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152651AF750
Description
PDE8A Polyclonal antibody specifically detects PDE8A in Human samples. It is validated for Western BlotSpecifications
PDE8A | |
Polyclonal | |
Western Blot | |
cAMP-specific cyclic nucleotide phosphodiesterase 8A, EC 3.1.4.17, FLJ16150, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, high-affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, HsT19550, phosphodiesterase 8A | |
The immunogen for this product maps to a region between residue 779 and 829 of human Phosphodiesterase 8A using the numbering given in entry NP_002596.1 (GeneID 5151). | |
0.1 mL | |
Lipid and Metabolism, Signal Transduction | |
5151 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction