Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582311 Supplier Invitrogen™ Supplier No. PA582311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTMGYQSGEL IGKELGEVPI NEKKADLLDT INSCIRIGKE WQGIYYAKKK NGDNIQQNVK IIPVIGQGGK IRHYVSIIRV CNGNNKAEKI SECVQSDTHT DNQTGKHKDR RKGSLDVKAV ASRATEVSSQ RRHSS Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 80%.

Phosphodiesterases (PDEs) regulate the intracellular levels of cAMP and cGMP. These cyclic nucleotides play an important role as second messengers in multiple physiologic processes, including regulation of vascular resistance, cardiac output, visceral motility, immune response, inflammation, neuroplasticity, vision, and reproduction. PDEs comprise a large superfamily of enzymes divided into 10 families. Different PDEs can be distinguished by their structure, tissue expression, localization, substrate specificity, regulation, and sensitivity to PDE inhibitors. Diversity in structure and specificity of function make PDEs promising targets for the pharmacotherapy of diseases modulated by cyclic nucleotide signaling (Hetman et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE8A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pde8a
Gene Accession No. O60658
Gene Alias AI551852; cAMP-specific cyclic nucleotide phosphodiesterase 8A; cAMP-specific cyclic nucleotide phosphodiesterase PDE8; FLJ16150; high affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8A; HsT19550; mmPDE8; Pde8; PDE8A; phosphodiesterase 8A; RNPDE8A
Gene Symbols Pde8a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDE8A. Recombinant protein control fragment (Product #RP-95154).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.