Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588405 Supplier Invitrogen™ Supplier No. PA588405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGCAPSIHIS ERLVAEDAPS PAAPPLSSGG PRLPQGQKTA ALPRTRGAGL LESELRDGSG KKVAVADVQF GPMRFHQDQL QVLLVFTKED NQCNGFCRAC EKAGFKCTVT KEAQAVLACF; Positive Samples: Mouse testis, Mouse eye, Mouse liver, Rat testis.

Phosphodiesterases (PDEs) regulate the intracellular levels of cAMP and cGMP. These cyclic nucleotides play an important role as second messengers in multiple physiologic processes, including regulation of vascular resistance, cardiac output, visceral motility, immune response, inflammation, neuroplasticity, vision, and reproduction. PDEs comprise a large superfamily of enzymes divided into 10 families. Different PDEs can be distinguished by their structure, tissue expression, localization, substrate specificity, regulation, and sensitivity to PDE inhibitors. Diversity in structure and specificity of function make PDEs promising targets for the pharmacotherapy of diseases modulated by cyclic nucleotide signaling (Hetman et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE8A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pde8a
Gene Accession No. O88502
Gene Alias AI551852; cAMP-specific cyclic nucleotide phosphodiesterase 8A; cAMP-specific cyclic nucleotide phosphodiesterase PDE8; FLJ16150; high affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8A; HsT19550; mmPDE8; Pde8; PDE8A; phosphodiesterase 8A; RNPDE8A
Gene Symbols Pde8a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human PDE8A (NP_0025961).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18584, 308776
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.