Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE8A Rabbit anti-Bovine, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502208417
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8417 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8417 Supplier Fabgennix Inc Supplier No. PD8A101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE8A Polyclonal antibody specifically detects PDE8A in Bovine, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen PDE8A
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Pde8a
Gene Accession No. O60658, O88502
Gene Alias AI551852; cAMP-specific cyclic nucleotide phosphodiesterase 8A; cAMP-specific cyclic nucleotide phosphodiesterase PDE8; FLJ16150; high affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8A; HsT19550; mmPDE8; Pde8; PDE8A; phosphodiesterase 8A; RNPDE8A
Host Species Rabbit
Immunogen Synthetic cyclic peptide from near the N terminus common to all PDE8A variants.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18584, 308776, 453612, 506787, 5151
Target Species Bovine, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.