Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE8B Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171817AF647
Description
PDE8B Polyclonal antibody specifically detects PDE8B in Human samples. It is validated for Western BlotSpecifications
PDE8B | |
Polyclonal | |
Western Blot | |
3'-5' cyclic nucleotide phosphodiesterase 8B, ADSD, Cell proliferation-inducing gene 22 protein, EC 3.1.4.17, FLJ11212, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8B, hsPDE8B, phosphodiesterase 8B, PPNAD3 | |
The immunogen for this product maps to a region between residue 835 and 885 of human Phosphodiesterase 8B using the numbering given in entry NP_003710.1 (GeneID 8622). | |
0.1 mL | |
Lipid and Metabolism, Signal Transduction | |
8622 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction