Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532981
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532981 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532981 Supplier Invitrogen™ Supplier No. PA532981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%); Dog, Horse (94%); Mouse, Rat, Bat, Elephant, Pig (88%); Opossum, Platypus (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Cyclic nucleotide phosphodiesterases (PDEs) catalyze hydrolysis of the 5-prime,3-prime-cyclic nucleotides cAMP and cGMP to the corresponding nucleoside 5-prime-monophosphates. Mammalian PDEs have been grouped into several families based on substrate affinities, inhibitor sensitivities, mode of regulation, and amino acid sequence homologies. The PDE8 family contains high-affinity cAMP-specific, IBMX (3-isobutyl-1-methyl-xanthine)-insensitive PDEs, such as PDE8B. All PDEs share a conserved C-terminal catalytic region and a variable N-terminal domain that presumably accounts for the distinctive regulatory properties unique to the individual families.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE8B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PDE8B
Gene Accession No. O95263
Gene Alias 3',5' cyclic nucleotide phosphodiesterase 8B; ADSD; B230331L10Rik; C030047E14Rik; cAMP-specific cyclic nucleotide phosphodiesterase PDE8; cell proliferation-inducing gene 22 protein; FLJ11212; High affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8B; hsPDE8B; Pde8b; phosphodiesterase 8B; PIG22; PPNAD3; RNPDE8B
Gene Symbols PDE8B
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human PDE8B.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8622
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.