Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE9A Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP100641G
Description
PDE9A Polyclonal specifically detects PDE9A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PDE9A | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, EC 3.1.4.1710phosphodiesterase PDE9A21, EC 3.1.4.35, FLJ90181, high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A, HSPDE9A2, phosphodiesterase 9A | |
Rabbit | |
68 kDa | |
0.05 mL | |
Lipid and Metabolism | |
5152 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PDE9A | |
Sequence from the C-terminal region of PDE9A | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction