Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE9A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572944
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572944 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572944 Supplier Invitrogen™ Supplier No. PA572944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE9A Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, IP

The protein encoded by this gene catalyzes the hydrolysis of cAMP and cGMP to their corresponding monophosphates. The encoded protein plays a role in signal transduction by regulating the intracellular concentration of these cyclic nucleotides. Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE9A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.05% sodium azide
Gene PDE9A
Gene Accession No. O70628, O76083, Q8QZV1
Gene Alias CGMP-specific 3',5'-cyclic phosphodiesterase type 9; FLJ90181; high affinity cGMP-specific 3',5'-cyclic phosphodiesterase 9A; HSPDE9A2; Pde8b; PDE9A; PDE9A1; phosphodiesterase 9A; phosphodiesterase PDE9A21
Gene Symbols PDE9A
Host Species Rabbit
Immunogen Sequence from the C-terminal region of PDE9A.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18585, 191569, 5152
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.