Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDF Antibody (OTI3F3), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273301AF532
Description
PDF Monoclonal specifically detects PDF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Full length recombinant protein of human PDF (NP_071736) produced in E.coli. | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI3F3 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
EC 3.5.1.88, GDF15, MIC1, PDF1A, peptide deformylase (mitochondrial), peptide deformylase, mitochondrial, peptide deformylase-like protein, PLAB, Polypeptide deformylase | |
Mouse | |
0.1 mL | |
64146 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction