Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDF Antibody (OTI3F3), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273301PECY55
Description
PDF Monoclonal specifically detects PDF in Human samples. It is validated for Flow Cytometry.Specifications
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
Full length recombinant protein of human PDF (NP_071736) produced in E.coli. | |
0.1 mL | |
64146.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI3F3 | |
Flow Cytometry | |
EC 3.5.1.88, GDF15, MIC1, PDF1A, peptide deformylase (mitochondrial), peptide deformylase, mitochondrial, peptide deformylase-like protein, PLAB, Polypeptide deformylase | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction