Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDF Monoclonal Antibody (OTI3C12)
GREENER_CHOICE

Catalog No. PIMA525563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525563 Supplier Thermo Scientific Supplier No. MA525563
Add to Cart
Add to Cart

PDF Monoclonal Antibody (OTI3C12)

PDF Monoclonal Antibody for Western Blot, ICC/IF, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDF
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PDF
Gene Accession No. Q9HBH1
Gene Alias 2610019N19Rik; PDF; PDF1A; peptide deformylase (mitochondrial); peptide deformylase, mitochondrial; peptide deformylase-like protein; peptide deformylase-like protein-like; polypeptide deformylase; similar to peptide deformylase-like protein
Gene Symbols PDF
Host Species Mouse
Immunogen Full length human recombinant protein of PDF produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64146
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.