Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDF Monoclonal Antibody (OTI3F3)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525562

Catalog No. PIMA525562


Only null left
Add to Cart

Description

Description

PDF Monoclonal Antibody for Western Blot, ICC/IF, Flow

Removes the formyl group from the N-terminal Met of newly synthesized proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PDF
Monoclonal
0.78 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HBH1
PDF
Full length human recombinant protein of PDF produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot, Immunocytochemistry
OTI3F3
Unconjugated
PDF
2610019N19Rik; PDF; PDF1A; peptide deformylase (mitochondrial); peptide deformylase, mitochondrial; peptide deformylase-like protein; peptide deformylase-like protein-like; polypeptide deformylase; similar to peptide deformylase-like protein
Mouse
Affinity Chromatography
RUO
64146
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.