Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564117 Supplier Invitrogen™ Supplier No. PA564117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESVAGFLACV PRFQAVQISG LDPNGEQVVW QASGWAARII QHEMDHLQGC LFIDKMDSRT FTNVYWMKVN D Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Removes the formyl group from the N-terminal Met of newly synthesized proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDF
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDF
Gene Accession No. Q9HBH1
Gene Alias 2610019N19Rik; PDF; PDF1A; peptide deformylase (mitochondrial); peptide deformylase, mitochondrial; peptide deformylase-like protein; peptide deformylase-like protein-like; polypeptide deformylase; similar to peptide deformylase-like protein
Gene Symbols PDF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDF. Recombinant protein control fragment (Product #RP-103512).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64146
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.