Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDF Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734573 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4573 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4573 Supplier Proteintech Group Inc Supplier No. 248421AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Removes the formyl group from the N-terminal Met of newly synthesized proteins.

Specifications

Antigen PDF
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PDF
Gene Accession No. Q9HBH1
Gene Alias PDF, PDF1A, Polypeptide deformylase
Gene Symbols PDF
Host Species Rabbit
Immunogen PDF Fusion Protein Ag18233
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64146
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.