Learn More
PDGF-BB Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00 - $2175.00
Specifications
Antigen | PDGF-BB |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P4750UG
![]() |
Invitrogen™
500P4750UG |
50 μg |
Each for $250.00
|
|
|||||
500P47100UG
![]() |
Invitrogen™
500P47100UG |
100 μg |
Each for $319.00
|
|
|||||
500-P47-1MG
![]() |
Invitrogen™
500P471MG |
1 mg |
Each for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPIFKKATVT LEDHLACKCE TVAAARPVT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PDGF-BB. Anti-Human PDGF-BB-specific antibody was purified by affinity chromatography employing an immobilized Human PDGF-BB matrix. Sandwich ELISA: To detect Human PDGF-BB by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human PDGF-BB (500-P47Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human PDGF-BB. Western Blot: To detect hPDGF-BB by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hPDGF-BB is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
PDGF-BB | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P01127 | |
5155 | |
E.coli-derived Recombinant Human PDGF-BB. | |
Antigen Affinity Chromatography | |
PDGFB |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
becaplermin; c-sis; FLJ12858; H-PDGF-BB; IBGC5; PDGF; pdgf protein; PDGF subunit B; PDGF, B chain; PDGF2; PDGF-2; PDGFB; PDGF-B; platelet derived growth factor subunit B; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor B; platelet-derived growth factor B chain; platelet-derived growth factor beta; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); proto-oncogene c-Sis; Sis; SSV | |
PDGFB | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.