Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF-C Polyclonal Antibody
PDGF-C Polyclonal Antibody
Supplier: Thermo Scientific PA547156
Description
PDGF-C Polyclonal Antibody for Western Blot, IHC (P), Neu

Specifications
PDGF-C | |
Polyclonal | |
Unconjugated | |
PDGFC | |
1110064L01Rik; AI647969; C76851; fallotein; MGC102297; Pdgfc; PDGF-C; PDGFC latent form; PDGFC receptor-binding form; platelet derived growth factor C; platelet-derived growth factor C; Platelet-derived growth factor C, latent form; Platelet-derived growth factor C, receptor-binding form; platelet-derived growth factor, C polypeptide; rScdfg; Scdgf; secretory growth factor-like protein; spinal cord-derived growth factor; UNQ174/PRO200; VEGF-E | |
Goat | |
Antigen affinity chromatography | |
RUO | |
56034 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9NRA1 | |
PDGFC | |
E. coli-derived recombinant human PDGF-C Gly230-Gly345. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction