Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDGF-C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547141
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547141 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547141 Supplier Invitrogen™ Supplier No. PA547141
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human (rh) PDGF-CC is observed and less than 1% cross-reactivity with rhPDGF-AA, rhPDGF-BB, rhPDGF-D, and recombinant rat PDGF-AB is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

Sphingosine Kinase 2 (Sphk2) catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).These receptors couple to multiple G proteins (e.g. S1P1 couples to Gi whereas S1P2 and S1P3 couple to Gq, G13 in addition to Gi) and regulate a extremely wide range of cellular events including cell motility, survival, apoptosis, migration and cellcell interaction. Important roles for S1P have also been reported in regulation of cardiogenesis, vascular maturation, oocyte survival, immune cell trafficking, cells of the neuronal system and bone cells. S1P levels are regulated by the activity of Sphk (Sphk1 and Sphk2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF-C
Applications Immunohistochemistry (Paraffin), Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PDGFC
Gene Accession No. Q8CI19
Gene Alias 1110064L01Rik; AI647969; C76851; fallotein; MGC102297; Pdgfc; PDGF-C; PDGFC latent form; PDGFC receptor-binding form; platelet derived growth factor C; platelet-derived growth factor C; Platelet-derived growth factor C, latent form; Platelet-derived growth factor C, receptor-binding form; platelet-derived growth factor, C polypeptide; rScdfg; Scdgf; secretory growth factor-like protein; spinal cord-derived growth factor; UNQ174/PRO200; VEGF-E
Gene Symbols PDGFC
Host Species Goat
Immunogen E. coli-derived recombinant mouse PDGF-CC Val235-Gly345.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54635
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.