Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF-C Rabbit anti-Human, Clone: 4, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | PDGF-C |
---|---|
Clone | 4 |
Dilution | ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
Applications | ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
PDGF-C Monoclonal specifically detects PDGF-C in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
PDGF-C | |
ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E | |
Recombinant Human PDGF-C Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
4 | |
ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
0.2 um filtered solution in PBS | |
56034 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title