Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PDGF R alpha Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. FAB9215G025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9215G025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9215G025 Supplier R&D Systems Supplier No. FAB9215G025UG
This item is not returnable. View return policy

Rat Monoclonal Antibody

PDGFR alpha Monoclonal specifically detects PDGFR alpha in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen PDGFR alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 189208
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.5 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P26618
Gene Alias alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PDGF R alpha Leu25-Glu524 (Asp65Glu, Gly439Ala, Thr440Ala), predicted Accession # P26618
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5156
Test Specificity Detects mouse PDGF R alpha in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.