Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Antibody (206), Allophycocyanin

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289543APC
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry.Specifications
PDGF R beta | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
206 | |
Flow Cytometry | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human PDGF R beta (NP_002600.1; Met1-Lys531). | |
0.1 mL | |
Oncogenes, Phospho Specific, Protein Kinase | |
5159 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction