Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Antibody (APB5), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143349AF488
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
PDGF R beta | |
Monoclonal | |
Alexa Fluor 488 | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
APB5 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
PDGFRB | |
The immunogen for this antibody was PDGF receptor beta. | |
0.1 mL | |
Phospho Specific, Protein Kinase | |
5159 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction