Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Antibody (RM0005-4C54), Alexa Fluor™ 532

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11061021AF532
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDGF R beta | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
PDGFRB | |
Mouse PDGF R beta (extracellular domain) | |
Primary | |
PDGF receptor beta (RM0005-4C54) | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0005-4C54 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Rat | |
0.1 mL | |
5159 | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction