Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Antibody (RM0005-4C54), DyLight 755

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11061021IR
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDGF R beta | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PDGFRB | |
Mouse PDGF R beta (extracellular domain) | |
0.1 mL | |
Oncogenes, Phospho Specific, Protein Kinase | |
5159 | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0005-4C54 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
PDGF receptor beta (RM0005-4C54) | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction