Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFA Antibody (MM0013-6D35), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060979AF532
Description
PDGFA Monoclonal specifically detects PDGFA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDGFA | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
PDGFA | |
Human recombinant PDGFA | |
0.1 mL | |
Primary | |
In western blots this antibody shown no cross-reactivity with E-Cadherin, and P-Cadherin. There is 25% crossreactivitywith rhPDGF-AB, and no cross-reactivity with rhPDGF-BB. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0013-6D35 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
PDGF-1, PDGF1PDGF A-chain, PDGF-A, Platelet-derived growth factor A chain, platelet-derived growth factor alpha chain, platelet-derived growth factor alpha isoform 2 preproprotein, platelet-derived growth factor alpha polypeptidePDGF subunit A, platelet-derived growth factor subunit A | |
Mouse | |
Protein G purified | |
Cell Cycle and Replication | |
5154.0 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction