Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFA Antibody (MM0013-6D35) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | PDGFA |
---|---|
Clone | MM0013-6D35 |
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017982
![]() |
Novus Biologicals
NB110609790.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11060979
![]() |
Novus Biologicals
NB11060979 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
PDGFA Monoclonal specifically detects PDGFA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDGFA | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
5154 | |
IgG2b | |
Protein G purified |
MM0013-6D35 | |
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
PDGF-1, PDGF1PDGF A-chain, PDGF-A, Platelet-derived growth factor A chain, platelet-derived growth factor alpha chain, platelet-derived growth factor alpha isoform 2 preproprotein, platelet-derived growth factor alpha polypeptidePDGF subunit A, platelet-derived growth factor subunit A | |
PDGFA | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title