Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFR alpha Antibody (16A1), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP144581AF488
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
PDGFR alpha | |
Monoclonal | |
Alexa Fluor 488 | |
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody 16A1 recognizes CD140a PDGF-RA, the 170 kDa alpha chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative or anti-proliferative roles in various tumours. HLDA VI.; WS Code E022 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
16A1 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
PDGFRA | |
CD140a-transfected NIH 3T3 cells | |
0.1 mL | |
Phospho Specific, Protein Kinase | |
5156 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction