Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFR alpha Antibody (16A1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP144581
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, In vitro assay, Flow (Cell Surface), CyTOF-ready.Specifications
PDGFR alpha | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000, In vitro assay, Flow (Cell Surface), CyTOF-ready | |
PDGFRA | |
CD140a-transfected NIH 3T3 cells | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody 16A1 recognizes CD140a PDGF-RA, the 170 kDa alpha chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative or anti-proliferative roles in various tumours. HLDA VI.; WS Code E022 | |
Store at 4C. Do not freeze. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, In vitro Assay, CyTOF | |
16A1 | |
Unconjugated | |
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
Mouse | |
170 kDa | |
0.1 mg | |
Protein Kinase | |
5156 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction