Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFR alpha Antibody (APA5), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143350IR
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
PDGFR alpha | |
Monoclonal | |
DyLight 755 | |
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG2a κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
APA5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
PDGFRA | |
The immunogen for this antibody was CD140a. | |
0.1 mL | |
Phospho Specific, Protein Kinase | |
5156 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction