Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR alpha Antibody (MM0004-8A89) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB017976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017976 0.025 mg
NB11060969 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017976 Supplier Novus Biologicals Supplier No. NB110609690.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PDGFR alpha
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone MM0004-8A89
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:100-400, Immunohistochemistry-Paraffin 1:100-1:400
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P16234
Gene Alias alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Gene Symbols PDGFRA
Host Species Mouse
Immunogen N-terminal recombinant human platelet derived growth factor receptor alpha fragment.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5156
Test Specificity PDGF Receptor alpha. No cross-reactivity is shown with hPDGF Receptor beta.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.