Learn More
Promega PDGFR alpha (D842V) Kinase Enzyme System, 10microg
Catalog No. PRV4480 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4480 10 μg
1 options
Catalog No. PRV4480 Supplier Promega Supplier No. V4480
Recombinant human PDGFR alpha (D842V) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR alpha is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.
  • Easily Screen and Profile PDGFRα (D842V) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C