Learn More
Promega PDGFR alpha Kinase Enzyme System, 1mg
Catalog No. PRV3722 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3722 1 mg
1 options
Catalog No. PRV3722 Supplier Promega Supplier No. V3722
Add to Cart
Add to Cart
Request a Quote
Recombinant human PDGFR alpha (amino acids 550-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR alpha is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.
  • Easily Screen and Profile PDGFRα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C