Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PDGF R alpha Antibody, R&D Systems™

Rat Monoclonal Antibody
$151.20 - $467.50
Specifications
| Antigen | PDGFR alpha |
|---|---|
| Clone | 1039504 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3221SP
![]() |
R&D Systems
MAB3221SP |
25 μg |
Each for $151.20
|
|
|||||
MAB3221100
![]() |
R&D Systems
MAB3221100 |
100 μg |
Each for $467.50
|
|
|||||
Description
PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| PDGFR alpha | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Mouse myeloma cell line NS0-derived mouse PDGF R alpha , Leu25-Glu524, Accession # P26618.3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1039504 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P26618.3 | |
| 5156 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title