Learn More
Promega PDGFR alpha (T674I) Kinase Enzyme System, 10microg
Catalog No. PRV4486 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4486 10 μg
1 options
Catalog No. PRV4486 Supplier Promega Supplier No. V4486
Recombinant human PDGFRα (Y674I) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile PDGFRα (T674I) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C