Learn More
Promega PDGFR alpha (T674I) Kinase Enzyme System, 1mg
Catalog No. PRV6594 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6594 1 mg
1 options
Catalog No. PRV6594 Supplier Promega Supplier No. V6594
Add to Cart
Add to Cart
Request a Quote
Recombinant human PDGFRα (Y674I) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile PDGFRα (T674I) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C