Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFRA Monoclonal Antibody (OTI4E7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807642

Description
PDGFRA (PDGFR-alpha, Platelet Derived Growth Factor Receptor alpha) belongs to the protein tyrosine kinase family, and binds to the alpha chain of PDGF. PDGFR exhibits two different isoforms, alpha and beta, which are coded by two different genes. PDGF-alpha receptors are expressed in oligodendrocyte progenitor cells while PDGF-beta receptors are expressed on neurons. PDGFR possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind and activate only PDGFR alpha, while PDGF BB can bind and activate both PDGFR alpha and beta. Members of the PDGF receptor subfamily are cell surface receptors that act as mitogens for cells of mesenchymal origin, and consist of alpha and beta isoforms of ∽190 and 125 kDa, respectively. Activated PDGF receptors act on signaling pathways that control cell growth, actin reorganization, migration, and differentiation. In humans, the gene is located on the q arm of chromosome 4. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.Specifications
PDGFRA | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P16234 | |
PDGFRA | |
Human recombit protein fragment corresponding to amino acids 789-1089 of human PDGFRA produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4E7 | |
Unconjugated | |
PDGFRA | |
CD140A, PDGFR-2, PDGFR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5156 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction