Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRA Mouse anti-Human, Clone: OTI1C10, lyophilized, TrueMAB™
SDP

Catalog No. 501668389
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8389 Supplier OriGene Supplier No. CF807645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

PDGFRA (PDGFR-alpha, Platelet Derived Growth Factor Receptor alpha) belongs to the protein tyrosine kinase family, and binds to the alpha chain of PDGF. PDGFR exhibits two different isoforms, alpha and beta, which are coded by two different genes. PDGF-alpha receptors are expressed in oligodendrocyte progenitor cells while PDGF-beta receptors are expressed on neurons. PDGFR possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind and activate only PDGFR alpha, while PDGF BB can bind and activate both PDGFR alpha and beta. Members of the PDGF receptor subfamily are cell surface receptors that act as mitogens for cells of mesenchymal origin, and consist of alpha and beta isoforms of ∽190 and 125 kDa, respectively. Activated PDGF receptors act on signaling pathways that control cell growth, actin reorganization, migration, and differentiation. In humans, the gene is located on the q arm of chromosome 4. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.

Specifications

Antigen PDGFRA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C10
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PDGFRA
Gene Accession No. P16234
Gene Alias CD140A, PDGFR-2, PDGFR2
Gene Symbols PDGFRA
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 789-1089 of human PDGFRA produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5156
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.