Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDGFRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565741 Supplier Invitrogen™ Supplier No. PA565741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGFVYLQPHSE HQGVVYCRAE AGGRSQISVK YQLLYVAVPS GPPSTTILAS SNKVKSGDDI SVLCTVLGEP DVEVEFTWIF PGQKDERPVT IQDTWRLIHR GLGHTTRISQ SVITVEDFET IDAGYYICTA QNLQGQTTV Highest antigen sequence identity to the following orthologs - mouse 93%, rat 94%.

PDGFRL bears significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in the PDGFRL gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and non-small cell lung cancers. This suggests PDGFRL may function as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGFRL
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDGFRL
Gene Accession No. Q15198
Gene Alias 1110039P19Rik; AV013190; PDGF receptor beta-like tumor suppressor; PDGFRL; PDGFR-like protein; PDGRL; platelet derived growth factor receptor like; platelet-derived growth factor receptor-like; platelet-derived growth factor receptor-like protein; platelet-derived growth factor-beta-like tumor suppressor; PRLTS
Gene Symbols PDGFRL
Host Species Rabbit
Immunogen Recombinant Human PDGFRL. Recombinant protein control fragment (Product #RP-106417).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5157
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.