Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDGFRL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106500
Description
Antibody detects endogenous levels of total PDGFRL.
PDGFRL bears significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in the PDGFRL gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and non-small cell lung cancers. This suggests PDGFRL may function as a tumor suppressor.
Specifications
| PDGFRL | |
| Polyclonal | |
| Unconjugated | |
| PDGFRL | |
| 1110039P19Rik; AV013190; PDGF receptor beta-like tumor suppressor; PDGFRL; PDGFR-like protein; PDGRL; platelet derived growth factor receptor like; platelet-derived growth factor receptor-like; platelet-derived growth factor receptor-like protein; platelet-derived growth factor-beta-like tumor suppressor; PRLTS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 290771, 5157, 68797 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15198, Q5RJP7, Q6PE55 | |
| PDGFRL | |
| A synthesized peptide derived from human PDGFRL(Accession Q15198), corresponding to amino acid residues A88-T138. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction