Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFRL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 162171AP150UL

Description
PDGFRL bears significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in the PDGFRL gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and non-small cell lung cancers. This suggests PDGFRL may function as a tumor suppressor.Specifications
PDGFRL | |
Polyclonal | |
Unconjugated | |
PDGFRL | |
PDGFR like protein, PDGFRL, PDGRL, PRLTS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290771, 5157, 68797 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15198, Q5RJP7, Q6PE55 | |
Pdgfrl | |
PDGFRL Fusion Protein Ag9218 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction