Learn More
Invitrogen™ PDH Antibody Cocktail
Antibody Cocktail
Supplier: Invitrogen™ 456799

Description
Anti-PDH Western Blot Kit; Western Blot kit (Cocktail of monoclonal antibodies) specific to Human, bovine PDH. This antibody is validated for use in Western Blot. C-V-OSCP (clone ) 4C11C10D12 - Antigen Human rec. mature OSCP Amount: 12.5 μg , Working concentration: 0.5 μg/mL PDH-E1-alpha (clone) 9H9AF5 - Human rec. PDH-E1-a Amount: 25 μg , Working concentration: 1 μg/mL PDH-E1-beta (clone) 17A5E2H8 - Porcine PDH Amount: 62.5 μg, Working concentration: 2.5 μg/mL PDH-E2 (clone) 15D3G9C11 - Porcine PDH Amount: 25 μg, Working concentration: 1 μg/mL PDH-E2/E3bp (clone) 13G2AE2BH5 - Porcine PDH Amount: 25 μg, Working concentration: 1 μg/mL.
Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation.
Specifications
PDH | |
Cocktail | |
1.5 mg/mL | |
HEPES buffered saline with 0.02% sodium azide; pH 7.5 | |
A7MB35, P08559, P11177, P11966, P26284, P29803, P35486, P35487, P49432, Q06437, Q9D051 | |
PDHA1, PDHA2, PDHB | |
IgG fraction | |
RUO | |
117098, 18597, 18598, 289950, 29554, 407109, 5160, 5161, 5162, 613610, 68263, 768012 | |
4°C, do not freeze | |
Liquid |
Western Blot, Immunocytochemistry | |
Cocktail | |
Unconjugated | |
PDHB | |
13G2AE2BH5; 15D3G9C11; 17A5E2H8; 2610103L06Rik; 4C11C10D12; 9H9AF5; AL024199; C81408; PDHA; PDHA1; Pdha-1; Pdha2; Pdha-2; PDHAD; PDHAL; PDHB; PDHBD; PDHCE1A; PDHE1-A type I; PDHE1-A type II; PDHE1-B; PHE1A; PHE1B; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase (lipoamide) alpha 2; pyruvate dehydrogenase (lipoamide) beta; pyruvate dehydrogenase alpha 1; pyruvate dehydrogenase alpha 2; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 alpha 1 subunit; pyruvate dehydrogenase E1 alpha 2; pyruvate dehydrogenase E1 alpha 2 subunit; pyruvate dehydrogenase E1 beta subunit; pyruvate dehydrogenase E1 component alpha subunit; pyruvate dehydrogenase E1 component alpha subunit precursor; PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT, TESTIS-SPECIFIC FORM, MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II); pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; Pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; pyruvate dehydrogenase E1 component subunit beta, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1 subunit alpha 1; pyruvate dehydrogenase E1 subunit alpha 2; pyruvate dehydrogenase E1 subunit beta; pyruvate dehydrogenase E1alpha subunit; pyruvate dehydrogenase, E1 beta polypeptide; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific; RP11-723P2.1; Unknown (protein for MGC:133847) | |
Mouse | |
150 μg | |
Primary | |
Human, Mouse, Rat, Bovine | |
Antibody | |
Ig |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.