Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDHA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595980 Supplier Invitrogen™ Supplier No. PA595980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FANDATFEIK KCDLHRLEEG PPVTTVLTRE DGLKYYRMMQ TVRRMELKAD QLYKQKIIRG FCHLCDGQEA CCVGLEAGIN PTDHLITAYR AHGFTFTRGL SVREILAELT GRKGGCAKGK GGSMHMYAKN FYGGNGIVGA QVPLGAGIAL ACKYNGKDEV CLTLYGDGAA NQGQIFEAYN MAALWKLPCI FICENNRYGM GTSVERAAAS TDYYKRGDFI PGLRVDGMDI LCVREATRFA AAYCRSGKGP ILMELQTYRY HGHSMSDPGV SYRTREEIQE VRSKSDPIML LKDRMVNSNL ASVEELKEID VEVRKEIEDA AQFATADPEP PLEELGYHIY SSDPPFEVRG ANQWIKFKSV S; Positive Samples: Mouse heart, Mouse skeletal muscle; Cellular Location: Mitochondrion matrix.

The pyruvate dehydrogenase (PDH) complex is a nuclear-encoded mitochondrial multienzyme complex that catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle. The PDH complex is composed of multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3). The E1 enzyme is a heterotetramer of two alpha and two beta subunits. This gene encodes the E1 alpha 1 subunit containing the E1 active site, and plays a key role in the function of the PDH complex. Mutations in this gene are associated with pyruvate dehydrogenase E1-alpha deficiency and X-linked Leigh syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDHA1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PDHA1
Gene Accession No. P08559, P26284, P35486
Gene Alias PDHA; PDHA1; Pdha-1; PDHAD; PDHCE1A; PDHE1-A type I; PHE1A; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase alpha 1; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 alpha 1 subunit; pyruvate dehydrogenase E1 component alpha subunit; pyruvate dehydrogenase E1 component alpha subunit precursor; pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1 subunit alpha 1; pyruvate dehydrogenase E1alpha subunit
Gene Symbols PDHA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-390 of human PDHA1 (NP_0002751).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18597, 29554, 5160
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.