Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDHA1A Polyclonal Antibody
PDHA1A Polyclonal Antibody
Supplier: Thermo Scientific PA572684
Description
PDHA1A Polyclonal Antibody for Western Blot

Specifications
PDHA1A | |
Polyclonal | |
Unconjugated | |
pdha1a | |
im:6895726; pdha1; pdha1 protein; pdha1a; pdha1b; PDHE1-A type I; PHE1A; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase (lipoamide) alpha 1a; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 alpha 1 subunit a; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 subunit alpha 1a; pyruvate dehydrogenase E1 subunit alpha 1a; wu:fd18b01; wu:fo96f03; zgc:73271; zgc:86692 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
406702 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.97 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
pdha1a | |
KLH conjugated synthetic peptide between 233~263 amino acids from the Central region of Zebrafish PDHA1. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction